Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 3, 4 | 676 | ||
GenBank mRNA | AK046669.1 | - | 3, 4 | 642 | ||
AK144950.1 | - | 1749 | ||||
AK164809.1 | - | 3, 4 | 642 | |||
AK166977.1 | - | 3, 4 | 669 | |||
BC031392.1 | - | 3, 4 | 620 | 4952407 |