Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lins | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191001.1 | NP_001177930.1 | 6 | 1539 | ||
NM_152815.2 | NP_690028.2 | 6 | 1554 | |||
GenBank mRNA | AB083158.1 | - | 4 | 1138 | ||
AK012554.1 | - | 6 | 1401 | |||
AK156066.1 | - | 6 | 1523 | |||
AK171170.1 | - | 6 | 1466 | |||
BC030943.1 | - | 8 | 1546 | |||
BC058648.1 | - | 6 | 1371 | |||
BC059228.1 | - | 1 | 540 | |||
BC138549.1 | - | 4 | 1212 | 8861569 |