Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153125.2 | NP_694765.2 | 19 | 5970 | ||
GenBank mRNA | AK122242.2 | - | 12 | 1407 | ||
BC026689.1 | - | 3 | 315 | |||
BC034649.1 | - | 9 | 1028 | |||
BC042603.1 | - | 13 | 1486 | |||
BC059898.1 | - | 10 | 1573 |