Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153128.2 | NP_694768.1 | 4 | 550 | ||
GenBank mRNA | AK034170.1 | - | 4 | 538 | ||
BC034514.1 | - | 4 | 550 | 3157064 |