Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153195.2 | NP_694875.2 | 2 | 384 | ||
GenBank mRNA | AK147797.1 | - | 2 | 336 | ||
AK147907.1 | - | 2 | 303 | |||
AK150190.1 | - | 2 | 159 | |||
AK150626.1 | - | 2 | 342 | |||
AK151025.1 | - | 2 | 342 | |||
AK152513.1 | - | 2 | 342 | |||
BC032153.1 | - | 2 | 326 | 5054087 | ||
CD563204.1 | - | 2 | 384 |