Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153388.4 | NP_700437.2 | 2 | 951 | ||
GenBank mRNA | AK081560.1 | - | 1 | 1234 | ||
AK163603.1 | - | 2 | 809 | |||
BC023036.1 | - | 1 | 564 | 5358628 | ||
BC023156.1 | - | 2 | 607 | 4951386 | ||
DQ078786.1 | - | 1 | 236 |