Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153585.5 | NP_705813.2 | 9 | 1162 | ||
GenBank mRNA | AK029103.1 | - | 9 | 1037 | ||
AK149944.1 | - | 9 | 995 | |||
AK157824.1 | - | 9 | 1061 | |||
AK171701.1 | - | 9 | 1142 | |||
BC025554.1 | - | 9 | 1023 | 5321869 | ||
BC036178.1 | - | 6 | 618 | |||
BC036179.1 | - | 9 | 1134 | 5059723 | ||
BC039183.1 | - | 9 | 1136 | 2631793 |