Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153587.2 | NP_705815.1 | 18 | 2922 | ||
GenBank mRNA | AK013999.1 | - | 1 | 277 | ||
AK030051.1 | - | 16 | 2668 | |||
AK032316.1 | - | 17 | 2820 | |||
AK160758.1 | - | 10 | 1686 | |||
AY341873.1 | - | 19 | 2911 | |||
AY341881.1 | - | 17 | 2716 | |||
BC035298.1 | - | 19 | 2922 | 4021865 | ||
BC038372.1 | - | 4 | 2827 |