Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153800.4 | NP_722495.3 | 4 | 457 | ||
GenBank mRNA | AK046097.1 | - | 4 | 459 | ||
AK078218.1 | - | 3 | 347 |