Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172288.2 | NP_758492.2 | 20 | 2901 | ||
GenBank mRNA | AK029317.1 | - | 20 | 2885 | ||
BC023915.1 | - | 21 | 2912 | 5323157 | ||
BC026845.1 | - | 20 | 2884 | 4235408 | ||
BC033476.1 | - | 20 | 2896 | |||
BC058707.1 | - | 1 | 1690 |