Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136471.1 | NP_001129943.1 | 5 | 577 | ||
NM_172297.1 | NP_758501.2 | 4 | 594 | |||
GenBank mRNA | AK134892.1 | - | 5 | 563 | ||
BC021915.1 | - | 4 | 594 | 4527178 |