Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002800.2 | NP_001002800.1 | 4 | 918 | ||
NM_001288753.1 | NP_001275682.1 | 4 | 843 | |||
NM_005496.3 | NP_005487.3 | 3 | 735 | |||
XM_006713459.3 | XP_006713522.1 | 4 | 688 | |||
XM_011512311.2 | XP_011510613.1 | 4 | 806 | |||
XM_011512312.2 | XP_011510614.1 | 3 | 427 | |||
XM_017005491.1 | XP_016860980.1 | 2 | 253 | |||
GenBank mRNA | AB019987.1 | - | 4 | 579 | ||
AF092564.1 | - | 2 | 209 | |||
AK002200.1 | - | 4 | 572 | |||
AK128143.1 | - | 4 | 497 | |||
AK225437.1 | - | 4 | 572 | |||
AK315070.1 | - | 4 | 572 | |||
AL136877.1 | - | 3 | 699 | |||
AL833949.1 | - | 3 | 1626 | |||
BC001557.1 | - | 4 | 554 | |||
BC065259.1 | - | 4 | 750 | |||
BC070161.1 | - | 4 | 572 | |||
BC106033.1 | - | 4 | 590 |