Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172455.3 | NP_766043.3 | 16 | 2100 | ||
GenBank mRNA | AK045663.1 | - | 14 | 1850 | ||
DQ061860.1 | - | 16 | 2101 |