Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172461.3 | NP_766049.2 | 6 | 860 | ||
GenBank mRNA | AK030042.1 | - | 6 | 819 | ||
AK043543.1 | - | 6 | 1099 | |||
AK054237.1 | - | 6 | 754 | |||
BC119588.1 | - | 5 | 694 | 40058889 | ||
BC119589.1 | - | 5 | 694 | 40058890 |