Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172465.4 | NP_766053.1 | 4 | 718 | ||
GenBank mRNA | AK032233.1 | - | 4 | 700 | ||
AK035745.1 | - | 3 | 2141 | |||
AK079248.1 | - | 4 | 685 | |||
BC042618.1 | - | 4 | 577 | 4950217 | ||
BC090832.1 | - | 4 | 536 | 6401763 |