Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172481.2 | NP_766069.1 | 4 | 620 | ||
GenBank mRNA | AK054426.1 | - | 4 | 621 | ||
AY355342.1 | - | 4 | 621 | |||
AY596198.1 | - | 2 | 456 | |||
BC066795.1 | - | 4 | 630 | 30473929 |