Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnptg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172529.3 | NP_766117.2 | 10 | 778 | ||
GenBank mRNA | AK044021.1 | - | 9 | 858 | ||
AK078230.1 | - | 807 | ||||
AY465529.1 | - | 10 | 786 | |||
BC023869.1 | - | 8 | 928 | |||
BC055872.1 | - | 10 | 778 | 4925332 |