Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172546.2 | NP_766134.1 | 13 | 3371 | ||
GenBank mRNA | AK033015.1 | - | 13 | 3372 | ||
AK149951.1 | - | 12 | 2669 | |||
AK172692.1 | - | 1 | 1148 | |||
AY151137.1 | - | 13 | 3333 | |||
BC024086.1 | - | 12 | 2719 | |||
BC031194.1 | - | 6 | 2478 | 5059479 |