Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172566.4 | NP_766154.2 | 1, 2 | 536 | ||
GenBank mRNA | AK042981.1 | - | 1 | 1436 | ||
AK086362.1 | - | 1, 2 | 537 | |||
BC119592.1 | - | 1, 2 | 537 | 40059040 |