Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199015.4 | NP_950180.3 | 9 | 819 | ||
GenBank mRNA | AK156583.1 | - | 9 | 800 | ||
BC110663.1 | - | 7 | 2438 | |||
BC145673.1 | - | 9 | 739 | 40130976 | ||
EU155107.1 | - | 6 | 430 |
Gene Symbol: Phf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172603.3 | NP_766191.1 | 7 | 642 | ||
GenBank mRNA | AK077176.1 | - | 7 | 637 | ||
BC116758.1 | - | 7 | 626 | 40126077 | ||
BC116760.1 | - | 7 | 626 | 40126079 | ||
EU155109.1 | - | 7 | 626 |
Gene Symbol: Phf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164327.1 | NP_001157799.1 | 7 | 621 | ||
GenBank mRNA | EU155108.1 | - | 7 | 621 |
Gene Symbol: Phf11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164289.1 | NP_001157761.1 | 8 | 1053 | ||
GenBank mRNA | EU155106.1 | - | 8 | 2923 |