Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172603.3 | NP_766191.1 | 4 | 376 | ||
GenBank mRNA | AK077176.1 | - | 4 | 371 | ||
BC116758.1 | - | 4 | 360 | 40126077 | ||
BC116760.1 | - | 4 | 360 | 40126079 | ||
EU155109.1 | - | 4 | 360 |
Gene Symbol: Gm6904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164329.1 | NP_001157801.1 | 4 | 376 |
Gene Symbol: Phf11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164327.1 | NP_001157799.1 | 4 | 355 | ||
GenBank mRNA | EU155108.1 | - | 4 | 355 |