Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172612.3 | NP_766200.1 | 1, 2 | 242 | ||
GenBank mRNA | AK039992.1 | - | 241 | |||
AK043639.1 | - | 1, 2 | 243 | |||
AK161870.1 | - | 1, 2 | 239 | |||
BC048531.1 | - | 1, 2 | 239 | 6535763 |