Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172678.3 | NP_766266.3 | 6 | 715 | ||
GenBank mRNA | AK028571.1 | - | 5 | 1429 | ||
AK031820.1 | - | 6 | 647 | |||
AK075984.1 | - | 6 | 634 | |||
AK145423.1 | - | 6 | 623 | |||
BC031137.1 | - | 6 | 651 | 4971848 | ||
BC032213.1 | - | 6 | 656 | 5323723 | ||
BC033277.1 | - | 6 | 636 | 4241041 |