Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289577.1 | NP_001276506.1 | 8 | 924 | ||
NM_001289578.1 | NP_001276507.1 | 7 | 708 | |||
NM_001289579.1 | NP_001276508.1 | 9 | 997 | |||
NM_001289580.1 | NP_001276509.1 | 9 | 1094 | |||
NM_172717.4 | NP_766305.2 | 8 | 924 | |||
GenBank mRNA | AK048930.1 | - | 9 | 1075 | ||
AK077629.1 | - | 8 | 896 | |||
AK133154.1 | - | 9 | 979 | |||
AK147892.1 | - | 7 | 683 | |||
AK148436.1 | - | 8 | 895 | |||
AK154038.1 | - | 8 | 906 | |||
AK155295.1 | - | 8 | 896 | |||
AK155525.1 | - | 8 | 896 | |||
AK169182.1 | - | 9 | 950 | |||
AK169193.1 | - | 8 | 879 | |||
BC049792.1 | - | 8 | 878 | 6510480 |