Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172735.2 | NP_766323.1 | 11 | 1764 | ||
GenBank mRNA | AK004251.1 | - | 1 | 748 | ||
AK088527.1 | - | 11 | 1765 | |||
AK146228.1 | - | 10 | 1735 | |||
AK151694.1 | - | 10 | 1716 | |||
AK167465.1 | - | 10 | 1721 | |||
AK167992.1 | - | 10 | 1735 | |||
AK168071.1 | - | 10 | 1730 | |||
AK209395.1 | - | 1 | 200 | |||
BC024560.1 | - | 4 | 736 | |||
BC037445.1 | - | 10 | 1674 | |||
BC050141.1 | - | 10 | 1713 | 30011212 |