Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172736.3 | NP_766324.1 | 10 | 1490 | ||
GenBank mRNA | AK034310.1 | - | 10 | 1495 | ||
AK170629.1 | - | 10 | 1498 | |||
BC034448.1 | - | 2 | 214 | |||
BC041775.1 | - | 10 | 1622 | |||
BC042658.1 | - | 10 | 1478 | 5365041 | ||
BC066768.1 | - | 10 | 1481 | 6407146 |