Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290477.1 | NP_001277406.1 | 6 | 1349 | ||
NM_001290478.1 | NP_001277407.1 | 6 | 1268 | |||
NM_172781.2 | NP_766369.2 | 6 | 1349 | |||
GenBank mRNA | AK044930.1 | - | 6 | 999 | ||
AK081448.1 | - | 6 | 1350 | |||
AK081937.1 | - | 6 | 1162 | |||
AK085099.1 | - | 6 | 1350 | |||
AK161786.1 | - | 6 | 1243 | |||
BC057137.1 | - | 6 | 1080 | |||
BC099954.1 | - | 6 | 1176 | 30081872 | ||
BC141251.1 | - | 6 | 1092 | 9055863 |