Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172816.3 | NP_766404.1 | 4, 5 | 900 | ||
GenBank mRNA | AK050482.1 | - | 4, 5 | 801 | ||
AK050494.1 | - | 4, 5 | 900 | |||
BC117544.1 | - | 4, 5 | 732 | 40105617 | ||
BC118521.1 | - | 4, 5 | 729 | 40105622 |