Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128922.1 | NP_001122394.1 | 3 | 2175 | ||
NM_005512.2 | NP_005503.1 | 3 | 2280 | |||
XM_005273903.4 | XP_005273960.2 | 3 | 2262 | |||
XM_011544935.2 | XP_011543237.1 | 3 | 2571 | |||
GenBank mRNA | BC052210.1 | - | 3 | 2172 | 4825609 | |
BC070079.1 | - | 3 | 2175 | 30344529 | ||
DA591993.1 | - | 1 | 72 | |||
Z24680.1 | - | 3 | 2131 |