Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172816.3 | NP_766404.1 | 6 | 1126 | ||
GenBank mRNA | AK050482.1 | - | 6 | 1027 | ||
AK050494.1 | - | 6 | 1126 | |||
BC117544.1 | - | 6 | 958 | 40105617 | ||
BC118521.1 | - | 6 | 955 | 40105622 |