Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP4K3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270425.1 | NP_001257354.1 | 31, 32 | 2811 | ||
NM_003618.3 | NP_003609.2 | 32, 33 | 2874 | |||
GenBank mRNA | AK021553.1 | - | 4, 5 | 378 | ||
AK291467.1 | - | 32, 33 | 2840 | |||
AK299889.1 | - | 24, 25 | 1903 | |||
AK314487.1 | - | 31, 32 | 2648 | |||
AK316144.1 | - | 5, 6 | 454 | |||
BC071579.1 | - | 31, 32 | 2803 | 5296205 |