Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291124.1 | NP_001278053.1 | 3 | 535 | ||
NM_173396.3 | NP_775572.1 | 3 | 535 | |||
GenBank mRNA | AK017884.1 | - | 3 | 403 | ||
AK029490.1 | - | 3 | 465 | |||
AK157623.1 | - | 3 | 443 | |||
AK178732.1 | - | 2 | 149 | |||
AK185386.1 | - | 2 | 169 | |||
AK207133.1 | - | 2 | 149 | |||
AK208156.1 | - | 2 | 149 | |||
AK219241.1 | - | 2 | 149 | |||
BC138705.1 | - | 3 | 448 | 8861727 | ||
BC138706.1 | - | 3 | 450 | 8861728 | ||
BC158022.1 | - | 2 | 297 | 9007328 | ||
CF731285.1 | - | 2 | 153 |