Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgif2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291124.1 | NP_001278053.1 | 4 | 1282 | ||
NM_173396.3 | NP_775572.1 | 3 | 1399 | |||
GenBank mRNA | AK017884.1 | - | 3 | 1261 | ||
AK029490.1 | - | 3 | 1329 | |||
AK157623.1 | - | 4 | 1190 | |||
AK182450.1 | - | 1 | 101 | |||
AK207161.1 | - | 1 | 101 | |||
AK209138.1 | - | 1 | 101 | |||
BC138705.1 | - | 3 | 1302 | 8861727 | ||
BC138706.1 | - | 3 | 1304 | 8861728 |