Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173413.3 | NP_775589.1 | 8 | 619 | ||
GenBank mRNA | AB232601.1 | - | 8 | 547 | ||
AK031251.1 | - | 8 | 566 | |||
AK084650.1 | - | 8 | 569 | |||
AK086085.1 | - | 3 | 161 | |||
AK155621.1 | - | 8 | 569 | |||
BC059208.1 | - | 8 | 601 | 6405462 | ||
BC157974.1 | - | 8 | 567 | 9007296 |