Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173413.3 | NP_775589.1 | 8 | 3954 | ||
GenBank mRNA | AK051218.1 | - | 6 | 1186 | ||
BC059208.1 | - | 8 | 3935 | 6405462 |