Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pskh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173432.2 | NP_775608.1 | 2 | 422 | ||
GenBank mRNA | AK053397.1 | - | 2 | 456 | ||
AK153861.1 | - | 2 | 469 | |||
AK154570.1 | - | 2 | 452 | |||
AK166338.1 | - | 2 | 445 | |||
AK178313.1 | - | 1 | 363 | |||
AK178805.1 | - | 1 | 363 | |||
AK214424.1 | - | 1 | 363 | |||
BC050128.1 | - | 2 | 422 | 30024345 |