Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291860.1 | NP_001278789.1 | 5 | 478 | ||
NM_005529.6 | NP_005520.4 | 5 | 478 | |||
XM_011541318.2 | XP_011539620.1 | 5 | 413 | |||
XM_017001120.1 | XP_016856609.1 | 5 | 554 | |||
XM_017001121.1 | XP_016856610.1 | 5 | 554 | |||
XM_017001122.1 | XP_016856611.1 | 5 | 554 | |||
GenBank mRNA | M85289.1 | - | 5 | 478 | ||
X62515.1 | - | 5 | 438 |