Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175001.3 | NP_778166.2 | 3 | 144 | ||
GenBank mRNA | AK077615.1 | - | 3 | 138 | ||
AK086913.1 | - | 3 | 123 | |||
AK164417.1 | - | 3 | 123 | |||
BC061191.1 | - | 3 | 134 | 6703228 | ||
BY790081.1 | - | 3 | 145 |