Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Synpo2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175132.3 | NP_780341.1 | 4 | 3836 | ||
GenBank mRNA | AK004253.1 | - | 1 | 761 | ||
AK053054.1 | - | 4 | 3836 | |||
AK084541.1 | - | 2 | 3123 |