Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177901.1 | NP_001171372.1 | 3 | 2008 | ||
NM_175311.4 | NP_780520.3 | 4 | 2021 | |||
GenBank mRNA | AK052459.1 | - | 3 | 1040 | ||
AK154195.1 | - | 3 | 2009 | |||
AK207186.1 | - | 1 | 138 | |||
BC058976.1 | - | 4 | 1951 | 5685033 |