Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175434.3 | NP_780643.2 | 4, 5 | 951 | ||
GenBank mRNA | AK046359.1 | - | 4, 5 | 931 | ||
AK083105.1 | - | 3, 4 | 706 | |||
BC115859.1 | - | 4, 5 | 797 | 40057292 | ||
BC115965.1 | - | 4, 5 | 797 | 40057290 |