Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175562.3 | NP_780771.1 | 2 | 656 | ||
GenBank mRNA | AB232635.1 | - | 2 | 539 | ||
AK041076.1 | - | 1 | 233 | |||
AK049038.1 | - | 2 | 656 | |||
AK053487.1 | - | 2 | 653 | |||
BC049787.1 | - | 2 | 636 | 6511404 |