Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176963.4 | NP_795937.1 | 5 | 908 | ||
GenBank mRNA | AK040998.1 | - | 5 | 909 | ||
AK158120.1 | - | 5 | 707 | |||
AK161687.1 | - | 5 | 707 | |||
AK170014.1 | - | 5 | 709 | |||
BC028818.1 | - | 5 | 711 | 5135570 |