Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177034.3 | NP_796008.2 | 9 | 2128 | ||
GenBank mRNA | AK032261.1 | - | 8 | 892 | ||
AK147583.1 | - | 9 | 2118 | |||
AK147623.1 | - | 9 | 2080 | |||
BC141181.1 | - | 9 | 1946 | 9055793 |