Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177103.4 | NP_796077.2 | 2 | 786 | ||
GenBank mRNA | AK028177.1 | - | 2 | 787 | ||
AK042087.1 | - | 2 | 678 | |||
AK043171.1 | - | 2 | 671 | |||
AK136120.1 | - | 1 | 378 | |||
AK158338.1 | - | 2 | 651 | |||
AY522918.1 | - | 2 | 650 | |||
BC067014.1 | - | 2 | 553 | 6812169 | ||
BC080830.1 | - | 2 | 670 | 30433573 |