Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177222.4 | NP_796196.3 | 14 | 1849 | ||
GenBank mRNA | AK132912.1 | - | 14 | 2073 | ||
AY423542.1 | - | 14 | 1780 | |||
AY485607.1 | - | 14 | 1849 | |||
AY485608.1 | - | 14 | 1849 | |||
BC072568.1 | - | 12 | 1691 | |||
BC120743.1 | - | 14 | 1753 | 40129666 | ||
BC125423.1 | - | 14 | 1753 | 40129938 | ||
BC144801.1 | - | 14 | 1753 |