Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177333.3 | NP_796307.2 | 11 | 1938 | ||
GenBank mRNA | AK088333.1 | - | 11 | 1932 | ||
AK089979.1 | - | 11 | 1948 | |||
AK131141.1 | - | 11 | 1922 | |||
AK141537.1 | - | 11 | 1938 | |||
BC031743.1 | - | 11 | 1926 | 4187527 |