Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177882.4 | NP_808550.1 | 3 | 263 | ||
GenBank mRNA | AK080438.1 | - | 3 | 263 | ||
BC138897.1 | - | 3 | 204 | 8861919 |