Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177900.4 | NP_808568.1 | 3 | 303 | ||
GenBank mRNA | AB107882.1 | - | 3 | 303 | ||
AK034300.1 | - | 3 | 312 | |||
AK158408.1 | - | 3 | 330 | |||
AY262758.1 | - | 3 | 312 | |||
AY269789.1 | - | 3 | 375 | |||
BC125339.1 | - | 3 | 290 | 40129854 | ||
BC125341.1 | - | 3 | 290 | 40129856 |