Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177911.4 | NP_808579.2 | 4 | 428 | ||
GenBank mRNA | AF486627.1 | - | 4 | 427 | ||
AK035279.1 | - | 4 | 429 | |||
BC141297.1 | - | 4 | 425 | 9055909 | ||
BC145622.1 | - | 4 | 425 | 9054190 |